Merck
  • Formaldehyde and hexamethylenetetramine as food additives: chemical interactions and toxicology.
Formaldehyde and hexamethylenetetramine as food additives: chemical interactions and toxicology.
Food additives and contaminants (1992-09-01)
P Restani, A R Restelli, C L Galli
ABSTRACT
Formaldehyde (FA) and hexamethylenetetramine (HMT) are used in cheese production to control gas-forming clostridia; FA also occurs naturally in some foods at levels of 1-20 mg/kg. The toxicology of FA and HMT are briefly discussed together with their reaction in foods. The most abundant end-product of FA in cheese is spinacine derived from the N-terminal histidine residue in gamma 2-casein. Acute and short term toxicological studies on spinacine enable a No Observed Effect Level of 300 mg/kg body weight/day to be determined, leading to an Acceptable Daily Intake (ADI) for man of 3 mg/kg body weight/day. From these data a Tolerance Level (TL) of 1800 mg spinacine/kg cheese can be derived, leading to a Safety Margin (SM) of 12.9. It is concluded that there is no appreciable health risk from consumption of cheese made using formaldehyde (Grana Padano) or hexamethylenetetramine (Provolone).
Sigma-Aldrich
Hexamethylenetetramine, puriss. p.a., reag. Ph. Eur., ≥99.5% (calc. to the dried substance)
Sigma-Aldrich
Hexamethylenetetramine, ACS reagent, ≥99.0%
Sigma-Aldrich
Hexamethylenetetramine, meets analytical specification of Ph. Eur., BP, 99-100.5% (calc. to the dried substance)
Methenamine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hexamethylenetetramine, ReagentPlus®, 99%