Merck
  • Lidocaine-prilocaine cream versus tetracaine gel for procedural pain in children.
Lidocaine-prilocaine cream versus tetracaine gel for procedural pain in children.
The Annals of pharmacotherapy (2002-03-29)
Anna Taddio, Mona G Y Gurguis, Gideon Koren
ABSTRACT
To determine the relative efficacy of lidocaine-prilocaine and tetracaine for procedural pain in children. Systematic review was performed (MEDLINE1990-June 2001) of all studies comparing the efficacy of these 2 modalities of pain relief in children undergoing painful cutaneous procedures. Search terms included lidocaine, prilocaine, tetracaine, and anesthesia. Eight studies compared lidocaine-prilocaine with tetracaine in children for 4 different procedures: intravenous cannulation, venipuncture, Port-a-Cath puncture, and laser therapy. When used as labeled (60 min for lidocaine-prilocaine, 30 min for tetracaine), the 2 modalities provided similar analgesic efficacy. When both anesthetics were applied for a similar duration of time (40 min, 60 min, 2 h), tetracaine provided superior anesthesia. Tetracaine was commonly associated with erythema, and lidocaine-prilocaine was associated with blanching of the skin. Lidocaine-prilocaine and tetracaine appear to be comparable for procedural pain relief when used as recommended. Tetracaine is more efficacious than lidocaine-prilocaine when both anesthetics are applied for the same amount of time.