Merck
Estimation of human exposure to styrene and ethylbenzene.
Toxicology (2000-04-27)
W Tang, I Hemm, G Eisenbrand
ABSTRACT
In the present studies, human exposure to styrene and to ethylbenzene (EB) is assessed on the basis of literature data. Total styrene and total EB exposure result from inhalation and from food intake. Styrene and EB inhaled represent the greatest proportion of the total intake. Styrene and EB content in food is mainly caused by migration from polymer packaging material. The daily styrene exposure is estimated to range from 18.2 to 55.2 microg/person, corresponding to an annual exposure of 6.7 to 20.2 mg/person. The daily EB exposure is estimated to be about 130 microg/person, corresponding to an annual exposure of 46 mg/person. Cigarette smoking is another important factor for styrene and EB intake by smokers.
Sigma-Aldrich
Ethylbenzene solution, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 10 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 8 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, 5% in chloroform-d (99.8 atom % D), TMS 2 %, NMR tube size 5 mm × 8 in.
Supelco
Ethylbenzene solution, 0.1% in chloroform-d (99.8 atom % D), chromium(III) acetylacetonate 0.1 mg/mL, TMS 0.01 %, NMR tube size 5 mm × 8 in.
Supelco
Ethylbenzene solution, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, 5% in chloroform-d (99.8 atom % D), TMS 1 %, NMR tube size 5 mm × 7 in.
Sigma-Aldrich
Ethylbenzene solution, 10% in chloroform-d (99.8 atom % D)
Supelco
Ethylbenzene solution, 10% in chloroform-d (99.8 atom % D), NMR tube size 5 mm × 8 in. , ultra-thin wall
Sigma-Aldrich
Ethylbenzene solution, 10% in chloroform-d (99.8 atom % D), NMR tube size 10 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, 0.1% in chloroform-d (99.8 atom % D), TMS 0.1 %
Sigma-Aldrich
Ethylbenzene solution, 0.1% in chloroform-d ("100%", 99.96 atom % D)
Sigma-Aldrich
Ethylbenzene solution, 1% in chloroform-d (99.8 atom % D)
Sigma-Aldrich
Ethylbenzene solution, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in. , ultra-thin wall
Sigma-Aldrich
Ethylbenzene, anhydrous, 99.8%
Supelco
Ethylbenzene, analytical standard
Supelco
Ethylbenzene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylbenzene, ReagentPlus®, 99%