Merck
  • Allergic contact dermatitis to benzyl alcohol in a hearing aid impression material.
Allergic contact dermatitis to benzyl alcohol in a hearing aid impression material.
American journal of contact dermatitis : official journal of the American Contact Dermatitis Society (1999-12-14)
D W Shaw
ABSTRACT
A patient with allergic contact dermatitis caused by benzyl alcohol in a hearing aid impression material and in topical medications is described. In addition, the patient had topical and probably systemic corticosteroid allergy. Benzyl alcohol allergy is reviewed. allergic contact dermatitis is the most commonly reported allergic reaction to benzyl alcohol. There is also 1 report of contact urticaria. Balsam of Peru patch tests are frequently positive. Reported allergic reactions to injected benzyl alcohol include generalized urticarial reactions, 1 generalized maculopapular reaction and 1 delayed localized reaction.
Product Description
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Supelco
Benzyl alcohol, analytical standard
Supelco
Benzyl alcohol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
USP
Benzyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%
Sigma-Aldrich
Benzyl alcohol, puriss., meets analytical specification of Ph. Eur., BP, NF, 99-100.5% (GC)
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard