Merck
  • Poly(4-vinylpyridine) supported acidic ionic liquid: a novel solid catalyst for the efficient synthesis of 2,3-dihydroquinazolin-4(1H)-ones under ultrasonic irradiation.
Poly(4-vinylpyridine) supported acidic ionic liquid: a novel solid catalyst for the efficient synthesis of 2,3-dihydroquinazolin-4(1H)-ones under ultrasonic irradiation.
Ultrasonics sonochemistry (2013-06-12)
Junke Wang, Yingxiao Zong, Rugang Fu, Yuying Niu, Guoren Yue, Zhengjun Quan, Xicun Wang, Yi Pan
ABSTRACT
A novel poly(4-vinylpyridine) supported acidic ionic liquid catalyst was synthesized by the reaction of 4-vinylpyridine with 1,3-propanesultone, followed by the polymerization and the addition of the heteropolyacid. Due to the combination of polymer features and ionic liquid, it acted as a heterogeneous catalyst to effectively catalyze the cyclocondensation reaction of anthranilamide with aldehydes under ultrasonic irradiation and afforded the corresponding 2,3-dihydro-4(1H)-quinazolinones compounds in good to excellent yields. In addition, the catalyst could be easily recovered by the filtration and reused six times without significant loss of catalytic activity. More importantly, the use of ultrasonic irradiation can obviously accelerate the reaction.
Sigma-Aldrich
1,3-Propanesultone, ≥99%