Merck
  • A fresh liver agar substrate for rearing small numbers of forensically important blow flies (Diptera: Calliphoridae).
A fresh liver agar substrate for rearing small numbers of forensically important blow flies (Diptera: Calliphoridae).
Journal of medical entomology (2014-06-06)
Susan V Gruner, Daniel H Slone
ABSTRACT
Forensically important calliphorids can be reared on a mixture of beef liver and agar. Small pieces of meat, especially fresh or frozen beef liver, will desiccate in 2-6 h, but this simple-to-make feeding substrate remains moist for at least 12 h at 25 and 30 degrees C without desiccation, even in small (5 g) amounts. We determined the survivorship of small numbers of Chrysomya megacephala (F.) (first-instar larvae to adult eclosion) raised on 5 g of liver agar and fresh beef liver. We found that all larvae raised on 5 g of liver died due to desiccation, but survivorship on 5 g of liver agar was equivalent to that on larger (50 g) pieces of either liver agar or beef liver.
Product Description
Millipore
Agar, suitable for microbiology
Millipore
Agar, suitable for microbiology
Sigma-Aldrich
Select agar
Sigma-Aldrich
Agar, Bacteriological, microbiology tested, suitable for plant cell culture, granular
Sigma-Aldrich
Agar, microbiology tested, suitable for plant cell culture, suitable for cell culture, powder
Sigma-Aldrich
Agar, suitable for plant cell culture
Sigma-Aldrich
Agar, ash 2.0-4.5%
Sigma-Aldrich
Noble agar
Sigma-Aldrich
Agar, High gel strength, suitable for plant cell culture
Sigma-Aldrich
Agar, Type A, suitable for plant cell culture
Sigma-Aldrich
Agar, Type M, suitable for plant cell culture
Sigma-Aldrich
Agar, suitable for plant cell culture, powder
Millipore
Agar, high purity, low ionic content, low gel strenght, suitable for microbiology
Millipore
Agar, powder, suitable for microbiology
Sigma-Aldrich
Bacteriological agar, for molecular biology