Merck
Vitamin A and dendritic cell differentiation.
Immunology (2014-05-21)
Marieke R Beijer, Georg Kraal, Joke M M den Haan
ABSTRACT
Vitamin A and its active metabolite retinoic acid are essential for the development and function of many tissues including the immune system. The induction of mucosal homing receptors on T and B cells by mucosal dendritic cells (DC) depends on the presence of vitamin A. Recent studies indicate that also the differentiation of CD11b+ DC subsets in the mucosa as well as the spleen depend on vitamin A signalling. As CD11b+ DC subsets exert non-redundant functions in anti-bacterial and anti-fungal immune responses, defects in CD11b+ DC differentiation will contribute to the clinical problems observed during vitamin A deficiency.
Sigma-Aldrich
Retinol, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)
Sigma-Aldrich
Retinol, BioXtra, ≥97.5% (HPLC), ~3100 U/mg
Sigma-Aldrich
Retinol, ≥95.0% (HPLC), ~2700 U/mg