Merck
  • Chronic effects of environmentally-relevant concentrations of lead in Pelophylax nigromaculata tadpoles: Threshold dose and adverse effects.
Chronic effects of environmentally-relevant concentrations of lead in Pelophylax nigromaculata tadpoles: Threshold dose and adverse effects.
Ecotoxicology and environmental safety (2014-04-15)
Min-Yi Huang, Ren-Yan Duan, Xiang Ji
ABSTRACT
Lead (Pb) is a common heavy metal in the natural environment, but its concentration has been increasing alongside widespread industrial and agricultural development in China. The dark-spotted frog Pelophylax (formerly Rana) nigromaculata (Anura: Ranidae) is distributed across East Asia and inhabits anthropogenic habitats such as farmland. Here, P. nigromaculata tadpoles (Gosner stage 19-46) were exposed to Pb at different concentrations (0, 40, 80, 160, 320, 640 and 1280µg/L) and Pb-induced survival, metamorphosis time, development, malformations, mobility and gonad structure were monitored. The results showed that above the threshold concentration of Pb, adverse effects were obvious. As the concentration of Pb increased, the adverse effects on different traits followed different patterns: the effects on hindlimb length, survival rate, metamorphosis rate, total malformation rate, swimming speed and jumping speed largely exhibited a linear pattern; the effects on snout-vent length, body mass and forelimb length largely exhibited a bimodal pattern. Sex ratio and gonadal histology were not affected by Pb, suggesting that Pb is not strongly estrogenic in P. nigromaculata.
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Lead, foil, 10mm disks, thickness 0.55mm, as rolled, 100%
Lead, foil, 8mm disks, thickness 1.0mm, as rolled, 99.99+%
Lead, foil, 10m coil, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 10mm disks, thickness 0.25mm, as rolled, 99.95%
Lead, foil, 15mm disks, thickness 0.5mm, as rolled, 99.99+%
Lead, foil, 10mm disks, thickness 0.04mm, 99.99+%
Lead, foil, 5m coil, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 1m coil, thickness 1.0mm, as rolled, 99.95%
Lead, foil, 6mm disks, thickness 0.025mm, 99.99+%
Lead, foil, 0.5m coil, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 6mm disks, thickness 0.01mm, 99.99+%
Lead, foil, 8mm disks, thickness 0.015mm, 99.99+%
Lead, foil, 10mm disks, thickness 0.008mm, 99.99+%
Lead, foil, 15mm disks, thickness 0.05mm, 99.99+%
Lead, wire reel, 2m, diameter 0.25mm, as drawn, 99.95%
Lead, wire reel, 2m, diameter 2.0mm, as drawn, 99.99+%
Lead, foil, not light tested, 25x25mm, thickness 0.0125mm, 99.99+%
Lead, foil, 2m coil, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 2m coil, thickness 0.12mm, coil width 25.4mm, as rolled, 99.95%
Lead, foil, 2m coil, thickness 0.25mm, as rolled, 99.95%
Lead, foil, 2m coil, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 25mm disks, thickness 0.25mm, as rolled, 99.99+%
Lead, foil, 25mm disks, thickness 0.55mm, as rolled, 100%
Lead, foil, 25mm disks, thickness 0.25mm, as rolled, 99.95%
Lead, foil, 25mm disks, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 2m coil, thickness 1.0mm, as rolled, 99.95%
Lead, foil, light tested, 50x50mm, thickness 0.01mm, 99.99+%