Merck
  • Comparison of virtual and manual tooth setups with digital and plaster models in extraction cases.
Comparison of virtual and manual tooth setups with digital and plaster models in extraction cases.
American journal of orthodontics and dentofacial orthopedics : official publication of the American Association of Orthodontists, its constituent societies, and the American Board of Orthodontics (2014-04-08)
Joon Im, Jung-Yul Cha, Kee-Joon Lee, Hyung-Seog Yu, Chung-Ju Hwang
ABSTRACT
The aim of this study was to compare the virtual and manual tooth setups with digital and plaster models in extraction cases by measuring various occlusal parameters and applying the American Board of Orthodontics objective grading system. Linear intra-arch and interarch dimensions (arch width and length, perimeter, overjet, and overbite), angular variables (tip, torque, and rotation), and American Board of Orthodontics objective grading system scores obtained from a digital virtual setup model were compared with those from a plaster model setup. The digital virtual setup model resulted in smaller arch perimeters than did the plaster setup model by 2.20 mm in the maxillary arch (P <0.01) and 1.30 mm in the mandibular arch (P <0.05). The digital virtual setup also exhibited significantly lower values for overbite and overjet (P <0.01). The digital virtual setup had tendencies toward mesial angulation of the anterior teeth, labial inclination of the maxillary anterior teeth (P <0.05), and distal in-rotation of the mandibular teeth (P <0.05). The resulting American Board of Orthodontics objective grading system evaluation showed that larger deductions for overjet, occlusal contact, and total score (P <0.01) were required for the digital than for the manual setup model. Digital and manual setups lead to similar measurements for intra-arch and interarch occlusal variables. However, because of the possibility of collision on proximal and occlusal contact, delicate adjustments in proximal and occlusal contacts are required.
Sigma-Aldrich
Drierite, with indicator, 6 mesh
Sigma-Aldrich
Drierite, with indicator, 10-20 mesh
Sigma-Aldrich
Calcium sulfate hemihydrate, purum, ≥97.0%
Sigma-Aldrich
Drierite, without indicator, 8 mesh
Sigma-Aldrich
Drierite, with indicator, 4 mesh
Sigma-Aldrich
Drierite, with indicator, 8 mesh
Sigma-Aldrich
Drierite, without indicator, 4 mesh
Sigma-Aldrich
Calcium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium sulfate dihydrate, puriss., meets analytical specification of NF, E 516, 99.0-101.0% (based on anhydrous substance)
Sigma-Aldrich
Calcium sulfate, −325 mesh, 99%
Sigma-Aldrich
Calcium sulfate dihydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Calcium sulfate dihydrate, ACS reagent, 98%