Merck
  • Atenolol versus propranolol for the treatment of infantile hemangiomas: a randomized controlled study.
Atenolol versus propranolol for the treatment of infantile hemangiomas: a randomized controlled study.
Journal of the American Academy of Dermatology (2014-03-25)
Alvaro Ábarzúa-Araya, Cristián P Navarrete-Dechent, Felipe Heusser, Javiera Retamal, María Soledad Zegpi-Trueba
ABSTRACT
Infantile hemangiomas have a dramatic response to propranolol, a nonselective beta-blocker. However, this treatment is not risk-free and many patients are excluded because of respiratory comorbidities. Atenolol is a cardioselective beta-blocker that may have fewer adverse events. We sought to evaluate the effectiveness of atenolol against propranolol in a noninferiority trial. In all, 23 patients met the inclusion criteria and were randomized to receive either atenolol or propranolol. Thirteen patients were treated with atenolol and 10 with propranolol. Follow-up was made at baseline, 2 weeks, 4 weeks, and then monthly for 6 months. Patients treated with atenolol had a complete response of 53.8% and 60% with propranolol, respectively. These results were nonsignificant (P = .68). Relevant adverse events were not reported. The reduced number of patients could have influenced our results. Atenolol appears to be as effective as propranolol. We did not find significant differences between these results or any adverse events.
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
USP
Propranolol hydrochloride, United States Pharmacopeia (USP) Reference Standard
USP
Atenolol, United States Pharmacopeia (USP) Reference Standard
Supelco
Propranolol hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Atenolol, European Pharmacopoeia (EP) Reference Standard
Propranolol hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(±)-Propranolol hydrochloride, ≥99% (TLC), powder
Supelco
(±)-Propranolol hydrochloride, analytical standard