Merck
  • Accelerated orthodontic tooth movement following le fort I osteotomy in a rodent model.
Accelerated orthodontic tooth movement following le fort I osteotomy in a rodent model.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons (2014-03-19)
Hao Yuan, Xiaofang Zhu, Jingting Lu, Jiewen Dai, Bing Fang, Steve G F Shen
ABSTRACT
In surgery-first accelerated orthognathic surgery, the clinical phenomenon of accelerated orthodontic tooth movement after osteotomy is a benefit compared with the conventional approach. However, because much of the literature on this phenomenon is based on empirical evidence and case reports, experimental animal-based studies are needed to verify and quantify this acceleration effect. The purpose of this prospective experimental study was to identify whether osteotomy procedures increase tooth movement. Le Fort I osteotomies were performed on the left maxillas in 15 male adult Sprague-Dawley rats. After surgery, a continuous force of 0.5 N was placed on the maxillary left first molar to move the tooth mesially. Another 15 rats had no surgery and served as controls. On days 1, 14, and 28, digital caliper measurements were taken to record tooth movement. In the experimental group, the maxillary left first molars moved significantly more rapidly on days 14 and 28 (P < .05). Histologic findings showed more active alveolar bone remodeling. Le Fort I osteotomy significantly accelerated the rate of orthodontic tooth movement. Histologically, more active and extensive bone remodeling was observed after osteotomy.
Sigma-Aldrich
Nickel, foil, thickness 0.25 mm, 99.995% trace metals basis
Sigma-Aldrich
Nickel, powder, <1 μm, 99.8% trace metals basis
Sigma-Aldrich
Nickel, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Phosphatase, Acid from wheat germ, ≥0.4 unit/mg solid
Sigma-Aldrich
Nickel, foil, thickness 0.125 mm, ≥99.9%
Nickel, wire reel, 20m, diameter 0.25mm, hard, 99.98%
Sigma-Aldrich
Nickel, wire, diam. 0.25 mm, ≥99.9%
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Nickel, rod, 1000mm, diameter 9.5mm, 99%
Nickel, foil, 50x50mm, thickness 0.125mm, as rolled, 99.99+%
Nickel, foil, 0.5m coil, thickness 1.0mm, annealed, 99.98%
Nickel, foil, 4mm disks, thickness 0.05mm, annealed, 99%
Nickel, foil, 4mm disks, thickness 0.005mm, as rolled, 99.9%
Nickel, foil, 0.5m coil, thickness 0.0025mm, 99.95%
Nickel, foil, 10mm disks, thickness 0.01mm, 99.95%
Nickel, foil, not light tested, 50x50mm, thickness 0.004mm, hard, 99%
Nickel, foil, 10m coil, thickness 0.3mm, coil width 1mm, as rolled, 99%
Nickel, foil, 10m coil, thickness 0.15mm, coil width 12mm, annealed, 99%
Nickel, foil, 2m coil, thickness 0.20mm, annealed, 99%
Nickel, foil, 50x50mm, thickness 0.50mm, annealed, 99%
Nickel, tube, 500mm, outside diameter 2.0mm, inside diameter 1.6mm, wall thickness 0.2mm, hard, 99.5%
Nickel, foil, 25x25mm, thickness 2.0mm, as rolled, 99.99+%
Nickel, foil, 25x25mm, thickness 0.15mm, annealed, 99%
Nickel, wire reel, 10m, diameter 0.75mm, as drawn, 99%
Nickel, wire reel, 100m, diameter 0.75mm, as drawn, 99%
Nickel, foil, 4mm disks, thickness 0.01mm, 99.95%
Nickel, foil, 10mm disks, thickness 0.008mm, 99.95%
Nickel, foil, 0.5m coil, thickness 0.003mm, 99.95%
Nickel, foil, 4mm disks, thickness 0.0075mm, 99.9%
Nickel, foil, 10m coil, thickness 0.20mm, coil width 1mm, annealed, 99%