Merck
Trypanosoma cruzi chemical proteomics using immobilized benznidazole.
Experimental parasitology (2014-03-19)
Andrea Trochine, Guzmán Alvarez, Sandra Corre, Paula Faral-Tello, Rosario Durán, Carlos I Batthyany, Hugo Cerecetto, Mercedes González, Carlos Robello
ABSTRACT
Benznidazole (Bzn) is a nitroimidazole drug currently used as first line treatment against Chagas disease, a neglected tropical disease caused by the flagellated protozoan Trypanosoma cruzi. Although the drug has been used since the late 1960s, its mechanism of action is not fully understood. In an attempt to study Bzn mode of action, a structurally modified derivative of the drug was synthesized and immobilized into a solid matrix. This allowed enrichment of T. cruzi proteins capable of binding immobilized Bzn, which were subsequently analysed by mass spectrometry. The proteins identified as specific non-covalent Bzn interactors were a homologue of the bacterial YjeF proteins, a Sec23A orthologue and the aldo-ketoreductase family member TcAKR. TcAKR is closely related to other enzymes previously associated with Bzn reductive activation such as NTRI and TcOYE. Thus, our untargeted search for Bzn binding partners allowed us to encounter proteins that could be related to drug reductive activation and/or resistance mechanisms.
Sigma-Aldrich
Sepharose 4B, 45-165 μm bead diameter
Sepharose 4B, Cytiva 17-0120-01, pack of 1 L
Sigma-Aldrich
N-Benzyl-2-nitro-1H-imidazole-1-acetamide, 97%
Sigma-Aldrich
Sepharose 6B, 6% Beaded Agarose, 45-165 μm (wet), fractionation range 10,000-1,000,000 Da (dextrans)
Sigma-Aldrich
Agarose, Type IV, Special High EEO
Sigma-Aldrich
Agarose, High Gelling Temperature
Sepharose 6B, Cytiva 17-0110-01, pack of 1 L
Sigma-Aldrich
Agarose, Type II-A, Medium EEO
Sigma-Aldrich
Agarose, For pulsed field electrophoresis running gel
Sigma-Aldrich
Agarose, Special High EEO
Sigma-Aldrich
Agarose, Type I-A, low EEO
Sigma-Aldrich
Agarose, Type I, low EEO
Sigma-Aldrich
Agarose, for molecular biology
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, low EEO
Sigma-Aldrich
Agarose, Wide range, for molecular biology
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature
Sigma-Aldrich
Agarose, Low EEO
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature, molecular biology grade
Sigma-Aldrich
Agarose, Low EEO, for Immunoelectrophoresis
Sigma-Aldrich
Agarose, Medium EEO, for molecular biology
Sigma-Aldrich
Agarose, High EEO, for molecular biology
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, Wide range/Standard 3:1
Sigma-Aldrich
Agarose, High EEO