Merck
  • Thulium:yttrium-aluminum-garnet laser for en bloc resection of bladder cancer: clinical and histopathologic advantages.
Thulium:yttrium-aluminum-garnet laser for en bloc resection of bladder cancer: clinical and histopathologic advantages.
Urology (2014-02-20)
Giovanni Muto, Devis Collura, Alessandro Giacobbe, Leonardo D'Urso, Gian Luca Muto, Andrea Demarchi, Sergio Coverlizza, Emanuele Castelli
ABSTRACT
To determine whether thulium:yttrium-aluminum-garnet laser resection of bladder tumor (TmLRBT) may offer advantages over classic resection. From April 2011 to September 2012, 55 consecutive patients newly diagnosed with clinical stage ≤T2 bladder cancer were enrolled in a prospective study on TmLRBT. Neoplasm was removed en bloc in all cases. When the tumor size was >3 cm, it was necessary to incise longitudinally and/or across the lesion and the bladder wall at its the base into 2 or more parts. All cases of non-muscle-invasive bladder cancer underwent second look in 30-90 days. Pathology reported urothelial carcinoma with Ta low grade in 31 patients (56.4%), T1 high grade in 18 (32.7%), and T2 high grade in 6 (10.9%). Histopathologic evaluation showed that the bladder detrusor was provided in all cases. Hemostasis was excellent, and no postoperative hematuria was reported. In a case of T1 G3, endoscopic re-evaluation showed a focal infiltration of the bladder detrusor, so the patient underwent radical cystectomy. To date, with a mean follow-up of 16 months (range, 8-25), the recurrence rate in patients with superficial disease is 14.5%. All recurrences were outside the site of first resection, and there was no progression in tumor grade. TmLRBT is a simple method that seems to overcome the "incise and scatter" problem associated with traditional transurethral resection of bladder tumor. Our initial data on staging accuracy and reduction of the local recurrence rate are encouraging.
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Aluminum, rod, 200mm, diameter 12.7mm, 99%
Aluminum, tube, 500mm, outside diameter 0.55mm, inside diameter 0.4mm, wall thickness 0.075mm, as drawn, 99.5%
Aluminum, rod, 200mm, diameter 15.0mm, as drawn, 99.999%
Aluminum, tube, 200mm, outside diameter 0.30mm, inside diameter 0.2mm, wall thickness 0.05mm, as drawn, 99.5%
Aluminum, sphere, 100pcs, diameter 6.0mm, 99%
Aluminum, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.999%
Aluminum, wire reel, 100m, diameter 0.25mm, hard, 99.99+%
Aluminum, wire reel, 1000m, diameter 0.30mm, as drawn, 99.5%
Aluminum, wire reel, 100m, diameter 0.5mm, as drawn, 99.95+%
Aluminum, wire reel, 1000m, diameter 0.7mm, as drawn, 99.5%
Aluminum, wire reel, 100m, diameter 0.05mm, as drawn, 99.99+%
Aluminum, wire reel, 100m, diameter 0.063mm, annealed, 99.99%
Aluminum, wire reel, 100m, diameter 0.25mm, as drawn, 99.5%
Aluminum, honeycomb, 600x600mm, thickness 52.3mm, cell size 6.3mm, cell wall 0.064mm, core density 0.083g/cm3, phenolic adhesive (resin), facing skin glass fiber 0.75mm, 99.9%
Aluminum, foam, 150x150mm, thickness 6.35mm, bulk density 0.2g/cm3, porosity 0.93, 2 pores/cm, aluminum 6101 grade, 98.5%
Yttrium, wire reel, 0.05m, diameter 1.0mm, 99.9%
Aluminum, honeycomb, 600x600mm, thickness 5mm, cell size 13mm, cell wall 0.1mm, core density 0.062g/cm3, phenolic adhesive (resin), 99.5%
Aluminum, honeycomb, 600x600mm, thickness 10mm, cell size 6.3mm, cell wall 0.064mm, core density 0.083g/cm3, phenolic adhesive (resin), 99.5%
Yttrium, wire reel, 0.1m, diameter 0.5mm, 99.9%
Aluminum, foam, 150x150mm, thickness 6.35mm, bulk density 0.2g/cm3, porosity 0.93, 8 pores/cm, aluminum 6101 grade, 98.5%
Aluminum, honeycomb, 300x300mm, thickness 26.5mm, cell size 6.3mm, cell wall 0.064mm, core density 0.083g/cm3, phenolic adhesive (resin), facing skin aluminum 0.55mm, 99.9%
Aluminum, honeycomb, 600x600mm, thickness 5mm, cell size 3.2mm, cell wall 0.025mm, core density 0.072g/cm3, phenolic adhesive (resin), 99.5%
Aluminum, honeycomb, 600x600mm, thickness 20mm, cell size 3.2mm, cell wall 0.025mm, core density 0.072g/cm3, phenolic adhesive (resin), 99.5%
Aluminum, honeycomb, 300x300mm, thickness 52.3mm, cell size 6.3mm, cell wall 0.064mm, core density 0.083g/cm3, phenolic adhesive (resin), facing skin glass fiber 0.75mm, 99.9%