Merck
  • Diagnosis and management of oral leishmaniasis--case series and literature review.
Diagnosis and management of oral leishmaniasis--case series and literature review.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons (2014-02-01)
Chen Nadler, Claes D Enk, Gilead T Leon, Yuval Samuni, Alexander Maly, Rakefet Czerninski
ABSTRACT
The worldwide prevalence of leishmaniasis is increasing because of ecologic changes and increased medical profession awareness. Furthermore, solitary cases have been recently reported in Western countries. The authors describe the epidemiology, mode of transmission, and diagnosis of leishmaniasis and present 4 oral cases treated with systemic, localized, or combined therapy. The authors suggest that clinicians should maintain a high index of suspicion for atypical, resistant, oral and perioral lesions in individuals with a history of traveling in certain geographic regions. After diagnosis, treatment should be determined jointly by experts from the fields of oral and maxillofacial surgery, oral medicine, and dermatology based on leishmaniasis species and clinical presentation.
Sigma-Aldrich
Amphotericin B solubilized, powder, γ-irradiated, BioXtra, suitable for cell culture
Amphotericin B for microbiological assay, European Pharmacopoeia (EP) Reference Standard
Amphotericin B for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Amphotericin B from Streptomyces sp., BioReagent, suitable for cell culture, ~80% (HPLC)
Sigma-Aldrich
Amphotericin B from Streptomyces sp., ~80% (HPLC), powder
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture
Amphotericin B, European Pharmacopoeia (EP) Reference Standard