Merck
Use of SERMs for treatment in postmenopausal women.
The Journal of steroid biochemistry and molecular biology (2014-01-01)
Joann V Pinkerton, Semara Thomas
ABSTRACT
Selective estrogen receptor modulators (SERMs) are synthetic non-steroidal agents which have varying estrogen agonist and antagonist activities in different tissues, most likely due to the receptor conformation changes associated with that SERM's binding and the subsequent effect on transcription. Clinical trials aim to differentiate amongst SERMs on selected target tissues for use in postmenopausal women including effects on breast, bone, cardiovascular venous thrombosis risk, endometrium, vagina, vasomotor symptoms, and brain. This paper describes differences in clinical effects on selected target tissues of SERMs that are approved, discontinued or in development. FDA approved SERMs include tamoxifen and toremifene used for prevention and treatment of breast cancer, raloxifene approved for prevention and treatment of osteoporosis and prevention of invasive breast cancer, and ospemifene approved for treatment of dyspareunia from menopausal vaginal atrophy. The FDA approved first tissue selective estrogen complex (TSEC) a pairing of conjugated equine estrogens with the SERM, bazedoxifene. This pairing reduces the risk of endometrial hyperplasia that can occur with the estrogenic component of the TSEC without the need for a progestogen in women with a uterus. It also allows for the estrogenic benefits on relief of hot flashes and prevention of bone loss without stimulating the breast or the endometrium. In clinical practice, the tissue-selective actions of SERMs, alone or paired with estrogens, allow for individualization in meeting the treatment needs of postmenopausal women by providing targeted tissue effects. This article is part of a Special Issue entitled 'Menopause'.
Sigma-Aldrich
Raloxifene hydrochloride, solid
Sigma-Aldrich
Tamoxifen citrate salt, ≥99%
Tamoxifen citrate for performance test, European Pharmacopoeia (EP) Reference Standard
Raloxifene hydrochloride, European Pharmacopoeia (EP) Reference Standard
Raloxifene hydrochloride for peak identification, European Pharmacopoeia (EP) Reference Standard
Tamoxifen citrate, European Pharmacopoeia (EP) Reference Standard
Supelco
Tamoxifen, analytical standard
Sigma-Aldrich
Toremifene citrate salt, ≥98% (HPLC)
Sigma-Aldrich
Tamoxifen, ≥99%