Merck
  • Bioavailability of iron, zinc, folic acid, and vitamin A from fortified maize.
Bioavailability of iron, zinc, folic acid, and vitamin A from fortified maize.
Annals of the New York Academy of Sciences (2013-12-18)
Diego Moretti, Ralf Biebinger, Maaike J Bruins, Birgit Hoeft, Klaus Kraemer
ABSTRACT
Several strategies appear suitable to improve iron and zinc bioavailability from fortified maize, and fortification per se will increase the intake of bioavailable iron and zinc. Corn masa flour or whole maize should be fortified with sodium iron ethylenediaminetetraacetate (NaFeEDTA), ferrous fumarate, or ferrous sulfate, and degermed corn flour should be fortified with ferrous sulfate or ferrous fumarate. The choice of zinc fortificant appears to have a limited impact on zinc bioavailability. Phytic acid is a major inhibitor of both iron and zinc absorption. Degermination at the mill will reduce phytic acid content, and degermed maize appears to be a suitable vehicle for iron and zinc fortification. Enzymatic phytate degradation may be a suitable home-based technique to enhance the bioavailability of iron and zinc from fortified maize. Bioavailability experiments with low phytic acid-containing maize varieties have suggested an improved zinc bioavailability compared to wild-type counterparts. The bioavailability of folic acid from maize porridge was reported to be slightly higher than from baked wheat bread. The bioavailability of vitamin A provided as encapsulated retinyl esters is generally high and is typically not strongly influenced by the food matrix, but has not been fully investigated in maize.
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Retinol, ≥95.0% (HPLC), ~2700 U/mg
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Sigma-Aldrich
Retinol, BioXtra, ≥97.5% (HPLC), ~3100 U/mg
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn+2 in THF, highly reactive Rieke®metal
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Retinol, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)
Iron, foil, light tested, 25x25mm, thickness 0.015mm, 99.85%
Iron, foil, not light tested, 50x50mm, thickness 0.001mm, 99.85%
Iron, foil, not light tested, 100x100mm, thickness 0.006mm, 99.85%
Iron, foil, not light tested, 150x150mm, thickness 0.008mm, 99.85%
Iron, wire reel, 1m, diameter 0.025mm, as drawn, 99.99+%
Iron, wire reel, 50m, diameter 0.25mm, hard, 99.5%
Iron, wire reel, 1m, diameter 1.0mm, as drawn, 99.99+%
Iron, tube, 500mm, outside diameter 12.0mm, inside diameter 10mm, wall thickness 1.0mm, annealed, 99.5%
Iron, tube, 100mm, outside diameter 8.0mm, inside diameter 5mm, wall thickness 1.5mm, annealed, 99.5%
Iron, wire reel, 0.5m, diameter 0.25mm, hard, 99.99+%
Iron, wire reel, 5m, diameter 0.25mm, hard, 99.99+%
Iron, wire reel, 20m, diameter 0.25mm, hard, 99.5%