Merck
  • Suppression of abdominal aortic aneurysm formation by inhibition of prolyl hydroxylase domain protein through attenuation of inflammation and extracellular matrix disruption.
Suppression of abdominal aortic aneurysm formation by inhibition of prolyl hydroxylase domain protein through attenuation of inflammation and extracellular matrix disruption.
Clinical science (London, England : 1979) (2013-11-22)
Aya Watanabe, Toshihiro Ichiki, Chikahiro Sankoda, Yusuke Takahara, Jiro Ikeda, Eriko Inoue, Tomotake Tokunou, Shiro Kitamoto, Kenji Sunagawa
ABSTRACT
In the present study we sought to determine the effect of CoCl2, an inhibitor of PHD (prolyl hydroxylase domain protein), on the development of AAA (abdominal aortic aneurysm). AAA was induced in C57BL/6 mice by periaortic application of CaCl2 (AAA group). NaCl (0.9%)-treated mice were used as a sham control (SHAM group). Mice were treated with 0.05% CoCl2 in the drinking water (AAA/CoCl2 group). At 1 and 6 weeks after the operation, aortic tissue was excised for further examination. After 6 weeks of CaCl2 treatment, aortic diameter and macrophage infiltration into the aortic adventitia were increased in the AAA group compared with the SHAM group. Treatment with CoCl2 reduced the aneurysmal size and macrophage infiltration compared with the AAA group. Aortic expression of inflammatory cytokines and MCP-1 (monocyte chemoattractant protein-1) and the activities of MMP-9 (matrix metalloproteinase-9) and MMP-2 were enhanced in the AAA group and attenuated in the AAA/CoCl2 group. Expression of cytokines and the activities of MMPs were already increased after 1 week of CaCl2 treatment, but were suppressed by CoCl2 treatment in association with reduced NF-κB (nuclear factor κB) phosphorylation. Treatment with CoCl2 in mice prevented the development of CaCl2-induced AAA in association with reduced inflammation and ECM (extracellular matrix) disruption. The results of the present study suggest that PHD plays a critical role in the development of AAA and that there is a therapeutic potential for PHD inhibitors in the prevention of AAA development.
Sigma-Aldrich
Cobalt(II) chloride, 97%
Sigma-Aldrich
Cobalt(II) chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Cobalt, foil, 25x25mm, thickness 1.0mm, as rolled, 99.99+%
Cobalt, foil, not light tested, 50x50mm, thickness 0.025mm, 99.9%
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, ≥99.99%
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Calcium chloride dihydrate, BioUltra, Molecular Biology, ≥99.5% (KT)
Sigma-Aldrich
Calcium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Calcium chloride dihydrate, Molecular Biology, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, puriss., meets analytical specification of Ph. Eur., USP, FCC, E509, 99-103%, ≤0.0001% Al
Sigma-Aldrich
Calcium chloride dihydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Calcium chloride dihydrate, tested according to Ph. Eur.
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Cobalt(II) chloride, AnhydroBeads, −10 mesh, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Cobalt, wire reel, 0.5m, diameter 0.5mm, hard, 99.99+%
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Cobalt, wire reel, 2m, diameter 0.05mm, hard, 99.9%
Cobalt, foil, not light tested, 25x25mm, thickness 0.008mm, 99.9%
Sigma-Aldrich
Cobalt(II) chloride, purum p.a., anhydrous, ≥98.0% (KT)
Sigma-Aldrich
Calcium chloride dihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Cobalt, foil, 4mm disks, thickness 0.01mm, 99.9%