Merck
  • In vivo magnetic resonance imaging and spectroscopy identifies oncolytic adenovirus responders.
In vivo magnetic resonance imaging and spectroscopy identifies oncolytic adenovirus responders.
International journal of cancer (2013-11-20)
O Hemminki, R Immonen, J Närväinen, A Kipar, J Paasonen, K T Jokivarsi, H Yli-Ollila, P Soininen, K Partanen, T Joensuu, S Parvianen, S K Pesonen, A Koski, M Vähä-Koskela, V Cerullo, S Pesonen, O H Gröhn, A Hemminki
ABSTRACT
At present, it is not possible to reliably identify patients who will benefit from oncolytic virus treatments. Conventional modalities such as computed tomography (CT), which measure tumor size, are unreliable owing to inflammation-induced tumor swelling. We hypothesized that magnetic resonance imaging (MRI) and spectroscopy (MRS) might be useful in this regard. However, little previous data exist and neither oncolytic adenovirus nor immunocompetent models have been assessed by MRS. Here, we provide evidence that in T2-weighted MRI a hypointense core area, consistent with coagulative necrosis, develops in immunocompetent Syrian hamster carcinomas that respond to oncolytic adenovirus treatment. The same phenomenon was observed in a neuroblastoma patient while he responded to the treatment. With relapse at a later stage, however, the tumor of this patient became moderately hyperintense. We found that MRS of taurine, choline and unsaturated fatty acids can be useful early indicators of response and provide detailed information about tumor growth and degeneration. In hamsters, calprotectin-positive inflammatory cells (heterophils and macrophages) were found in abundance; particularly surrounding necrotic areas in carcinomas and T cells were significantly increased in sarcomas, when these had been treated with a granulocyte-macrophage colony-stimulating factor-producing virus, suggesting a possible link between oncolysis, necrosis (seen as a hypointense core in MRI) and/or immune response. Our study indicates that both MRI and MRS could be useful in the estimation of oncolytic adenovirus efficacy at early time points after treatment.
Sigma-Aldrich
Taurine, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Choline chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
Taurine, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
Taurine, ≥99%
Sigma-Aldrich
Choline chloride, ≥98%
Sigma-Aldrich
Choline chloride, ≥99%
Supelco
Taurine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Taurine, ≥98%, FG
SAFC
Taurine
Sigma-Aldrich
Choline chloride, BioUltra, ≥99.0% (AT)
Supelco
Choline chloride, Pharmaceutical Secondary Standard; Certified Reference Material