Merck
  • Decontamination of prions in a plasma product manufacturing environment.
Decontamination of prions in a plasma product manufacturing environment.
Transfusion (2013-09-17)
Anne Bellon, Emmanuel Comoy, Steve Simoneau, Sandrine Mornac, Capucine Dehen, Audrey Perrin, Aude Arzel, Samuel Arrabal, Henry Baron, Hubert Laude, Bruno You, Jean-Philippe Deslys, Benoit Flan
ABSTRACT
The high resistance of prions to inactivating treatments requires the proper management of decontaminating procedures of equipment in contact with materials of human or animal origin destined for medical purposes. Sodium hydroxide (NaOH) is widely used today for this purpose as it inactivates a wide variety of pathogens including prions. Several NaOH treatments were tested on prions bound to either stainless steel or chromatographic resins in industrial conditions with multiple prion strains. Data show a strong correlation between inactivation results obtained by immunochemical detection of the prion protein and those obtained with infectivity assays and establish effective inactivation treatments for prions bound to stainless steel or chromatographic resins (ion exchange and affinity), including treatments with lower NaOH concentrations. Furthermore, no obvious strain-specific behavior difference was observed between experimental models. The results generated by these investigations show that industrial NaOH decontamination regimens (in combination with the NaCl elution in the case of the chromatography process) attain substantial prion inactivation and/or removal between batches, thus providing added assurance to the biologic safety of the final plasma-derived medicinal products.
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Ascarite®, Sodium hydroxide-coated silica, 8-20 mesh
Sigma-Aldrich
Ascarite®, Sodium hydroxide-coated silica, 20-30 mesh