Merck
  • Gas chromatography coupled to mass spectrometry analysis of volatiles, sugars, organic acids and aminoacids in Valencia Late orange juice and reliability of the Automated Mass Spectral Deconvolution and Identification System for their automatic identification and quantification.
Gas chromatography coupled to mass spectrometry analysis of volatiles, sugars, organic acids and aminoacids in Valencia Late orange juice and reliability of the Automated Mass Spectral Deconvolution and Identification System for their automatic identification and quantification.
Journal of chromatography. A (2012-04-27)
Manuela Cerdán-Calero, José María Sendra, Enrique Sentandreu
ABSTRACT
Neutral volatiles and non-volatile polar compounds (sugars, organics acids and aminoacids) present in Valencia Late orange juice have been analysed by Gas Chromatography coupled to Mass Spectrometry (GC-MS). Before analysis, the neutral volatiles have been extracted by Headspace-Solid Phase Microextraction (HS-SPME), and the non-volatile polar compounds have been transformed to their corresponding volatile trimethylsilyl (TMS) derivatives. From the resulting raw GC-MS data files, the reliability of the Automated Mass Spectral Deconvolution and Identification System (AMDIS) to perform accurate identification and quantification of the compounds present in the sample has been tested. Hence, both raw GC-MS data files have been processed automatically by using AMDIS and manually by using Xcalibur™, the manufacturer's data processing software for the GC-MS platform used. Results indicate that the reliability of AMDIS for accurate identification and quantification of the compounds present in the sample strongly depends on a number of operational settings, for both the MS and AMDIS, which must be optimized for the particular type of assayed sample. After optimization of these settings, AMDIS and Xcalibur™ yield practically the same results. A total of 85 volatiles and 22 polar compounds have been identified and quantified in Valencia Late orange juice.
Product Description
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 24 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 23 ga
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with manual holder, needle size 23 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), metal alloy fiber, df 100 μm(PDMS, needle size 23 ga, for use with autosampler