Merck
  • Preparation of magnetic Fe3O4@SiO2 nanoparticles for immobilization of lipase.
Preparation of magnetic Fe3O4@SiO2 nanoparticles for immobilization of lipase.
Journal of nanoscience and nanotechnology (2014-04-17)
Wei Liu, Fang Zhou, Xiao-Yun Zhang, Yue Li, Xiang-Yu Wang, Xi-Ming Xu, Ye-Wang Zhang
ABSTRACT
Magnetic Fe3O4 nanoparticles were prepared with coprecipitatation method and covered with SiO2 to form the core-shell Fe3O4@SiO2 nanoparticles. Then the nanoparticles were modified with glutaradehyde for functionalization of the surface to aldehyde groups. The transmission electron microscopy confirmed the core-shell structure and revealed that the size of the nanoparticles was around 10 nm. It was observed that the lipase was immobilized on the nanoparticles successfully from the Fourier transform infrared spectra. The immobilized lipase on Fe3O4@SiO2 nanoparticles was characterized and compared to free enzyme. There are no significant differences observed in the optimal pH, temperature and Km before and after immobilization. However, the immobilized lipase displayed higher relative activity in the range of pH from 7.0 to 9.5. Compare with the free enzyme, the immobilized one showed higher thermal stability at temperature range from 30 to 70 degrees C, especially at high temperature. The relative activity of immobilized enzyme was 5.8 fold of the free lipase at 70 degrees C after 10 h incubation. Thus, the prepared lipase was proved to have the advantages like higher relative activity, better stability, broader pH range and easy to recovery. These results suggest that immobilization of lipase on Fe3O4@SiO2 nanoparticles has the potential industrial applications.
Sigma-Aldrich
Silica
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Silica, nanopowder, 99.8% trace metals basis
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from Aspergillus oryzae, solution, ≥100,000 U/g, white, beige
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Silica, mesostructured, MSU-F (cellular foam)
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g