Merck
  • Erythrocyte-derived microvesicles amplify systemic inflammation by thrombin-dependent activation of complement.
Erythrocyte-derived microvesicles amplify systemic inflammation by thrombin-dependent activation of complement.
Arteriosclerosis, thrombosis, and vascular biology (2013-12-07)
Daniel Zecher, Arun Cumpelik, Jürg A Schifferli
ABSTRACT
Transfusion of aged blood has been associated with increased morbidity and mortality in critically ill patients. During storage, erythrocytes release increasing numbers of microvesicles (red blood cell-derived microvesicles [RBC-MV]). We hypothesized that RBC-MV mediate some of the deleterious effects of aged blood transfusions. We established a murine transfusion model using RBC-MV purified from aged mouse erythrocytes. Injection of RBC-MV into healthy mice had no effect. However, they aggravated pulmonary leukocyte sequestration and peripheral blood leukopenia induced by lipopolysaccharides. Lipopolysaccharide-induced proinflammatory cytokines were significantly increased in plasma after RBC-MV injection. These effects were not seen in C5aR-deficient mice. In vitro, RBC-MV bound C3 fragments after incubation with plasma but failed to bind immunoglobulins, C1q, or mannose-binding lectin. Preventing thrombin generation inhibited complement activation in vitro and in vivo and reversed the proinflammatory effects of RBC-MV in lipopolysaccharide-primed mice. Finally, the RBC-MV-induced phenotype was recapitulated using phosphatidylserine-expressing liposomes, suggesting that surface expression of phosphatidylserine by RBC-MV was mechanistically involved. These results point toward a thrombin-dependent mechanism of complement activation by RBC-MV independent of the classical, lectin, or alternative pathway. Besides identifying RBC-MV as potential mediators of transfusion-related morbidity, our findings may be relevant for other inflammatory disorders involving intravascular microvesicle release, for example, sickle cell disease or thrombotic microangiopathy.
Sigma-Aldrich
Thrombin from bovine plasma, lyophilized powder, 40-500 NIH units/mg protein (biuret)
Sigma-Aldrich
Thrombin from human plasma, lyophilized powder, Suitable for routine use in the thrombin time test
Sigma-Aldrich
Thrombin from human plasma, lyophilized powder, ≥2800 NIH units/mg protein (E1%/280, 18.3)
Sigma-Aldrich
Thrombin from human plasma, lyophilized powder, ≥2,000 NIH units/mg protein (E1%/280, 18.3)
Sigma-Aldrich
Thrombin from human plasma, lyophilized powder, ≥1,000 NIH units/mg protein (E1%/280, 18.3)
Sigma-Aldrich
Thrombin human, BioUltra, recombinant, expressed in HEK 293 cells, aqueous solution, ≥95% (SDS-PAGE)