Merck
  • Regulation of renal phosphate transport by FGF23 is mediated by FGFR1 and FGFR4.
Regulation of renal phosphate transport by FGF23 is mediated by FGFR1 and FGFR4.
American journal of physiology. Renal physiology (2013-11-22)
Jyothsna Gattineni, Priyatharshini Alphonse, Qiuyu Zhang, Nisha Mathews, Carlton M Bates, Michel Baum
ABSTRACT
Fibroblast growth factor 23 (FGF23) is a bone-derived hormone that acts on the proximal tubule to decrease phosphate reabsorption and serum levels of 1,25-dihydroxyvitamin D₃ [1,25(OH)₂ Vitamin D₃]. Abnormal FGF23 metabolism has been implicated in several debilitating hypophosphatemic and hyperphosphatemic disorders. The renal receptors responsible for the phosphaturic actions of FGF23 have not been elucidated. There are four fibroblast growth factor receptors (FGFR); 1-4 with "b" and "c" isoforms for receptors 1, 2, and 3. FGFR1, 3, and 4 are expressed in the mouse proximal tubule, and deletion of any one receptor did not affect serum phosphate levels, suggesting that more than one receptor is involved in mediating the phosphaturic actions of FGF23. To determine the receptors responsible for the phosphaturic actions of FGF23, we studied Fgfr1 (kidney conditional) and Fgfr4 (global) double mutant mice (Fgfr1⁻/⁻/Fgfr4⁻/⁻). Fgfr1⁻/⁻/Fgfr4⁻/⁻ mice have higher FGF23 levels than their wild-type counterparts (108.1 ± 7.3 vs. 4,953.6 ± 675.0 pg/ml; P < 0.001). Despite the elevated FGF23 levels, Fgfr1⁻/⁻/Fgfr4⁻/⁻ mice have elevated serum phosphorus levels, increased brush-border membrane vesicle (BBMV) phosphate transport, and increased Na-P(i) cotransporter 2c (NaPi-2c) protein expression compared with wild-type mice. These data are consistent with FGFR1 and FGFR4 being the critical receptors for the phosphaturic actions of FGF23.
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-1, partially purified powder, ≥300,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-2, aqueous solution, ≥85,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-5, lyophilized powder, ≥400,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type HP-2S, aqueous solution, ≥90,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-3AF, aqueous solution, ≥60,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-3, aqueous solution, ≥90,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type HP-2, aqueous solution, ≥100,000 units/mL
Sigma-Aldrich
β-Glucuronidase from limpets (Patella vulgata), Type L-II, lyophilized powder, 1,000,000-3,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from limpets (Patella vulgata), aqueous solution, ≥85,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, ≥10,000,000 units/g protein (30 min assay), recombinant, expressed in E. coli overproducing strain, lyophilized powder
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, >20,000,000 units/g protein, recombinant, expressed in E. coli, aqueous glycerol solution
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, ≥20,000 units/mg protein, recombinant, expressed in E. coli overproducing strain, lyophilized powder
Sigma-Aldrich
β-Glucuronidase from Helix aspersa (garden snail), Type HA-4
Sigma-Aldrich
β-Glucuronidase from bovine liver, Type B-1, ≥1,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, Type IX-A, lyophilized powder, 1,000,000-5,000,000 units/g protein (30 min assay)
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, aqueous glycerol solution, ≥5,000,000 units/g protein, pH 6.8 (biuret)
Sigma-Aldrich
β-Glucuronidase from bovine liver, Type B-3, ≥2,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, Type VII-A, lyophilized powder, 5,000,000-20,000,000 units/g protein, pH 6.8 (30 min assay)