Merck
  • Anti-inflammatory and immunomodulatory properties of α1-antitrypsin without inhibition of elastase.
Anti-inflammatory and immunomodulatory properties of α1-antitrypsin without inhibition of elastase.
Proceedings of the National Academy of Sciences of the United States of America (2013-08-27)
Danny Jonigk, Mariam Al-Omari, Lavinia Maegel, Meike Müller, Nicole Izykowski, Jaewoo Hong, Kwangwon Hong, Soo-Hyun Kim, Martina Dorsch, Ravi Mahadeva, Florian Laenger, Hans Kreipe, Armin Braun, Galit Shahaf, Eli C Lewis, Tobias Welte, Charles A Dinarello, Sabina Janciauskiene
ABSTRACT
The rationale of α1-antitrypsin (AAT) augmentation therapy to treat progressive emphysema in AAT-deficient patients is based on inhibition of neutrophil elastase; however, the benefit of this treatment remains unclear. Here we show that clinical grade AAT (with elastase inhibitory activity) and a recombinant form of AAT (rAAT) without anti-elastase activity reduces lung inflammatory responses to LPS in elastase-deficient mice. WT and elastase-deficient mice treated with either native AAT or rAAT exhibited significant reductions in infiltrating neutrophils (23% and 68%), lavage fluid levels of TNF-α (70% and 80%), and the neutrophil chemokine KC (CXCL1) (64% and 90%), respectively. Lung parenchyma TNF-α, DNA damage-inducible transcript 3 and X-box binding protein-1 mRNA levels were reduced in both mouse strains treated with AAT; significantly lower levels of these genes, as well as IL-1β gene expression, were observed in lungs of AAT-deficient patients treated with AAT therapy compared with untreated patients. In vitro, LPS-induced cytokines from WT and elastase-deficient mouse neutrophils, as well as neutrophils of healthy humans, were similarly reduced by AAT or rAAT; human neutrophils adhering to endothelial cells were decreased by 60-80% (P < 0.001) with either AAT or rAAT. In mouse pancreatic islet macrophages, LPS-induced surface expression of MHC II, Toll-like receptor-2 and -4 were markedly lower (80%, P < 0.001) when exposed to either AAT or rAAT. Consistently, in vivo and in vitro, rAAT reduced inflammatory responses at concentrations 40- to 100-fold lower than native plasma-derived AAT. These data provide evidence that the anti-inflammatory and immunomodulatory properties of AAT can be independent of elastase inhibition.