Merck
  • Characterization of polysaccharides from Ganoderma spp. using saccharide mapping.
Characterization of polysaccharides from Ganoderma spp. using saccharide mapping.
Carbohydrate polymers (2013-08-06)
Ding-Tao Wu, Jing Xie, De-Jun Hu, Jing Zhao, Shao-Ping Li
ABSTRACT
Polysaccharides from Ganoderma spp. and their adulterants were firstly investigated and compared using saccharide mapping, enzymatic (endo-1,3-β-D-glucanase and pectinase) digestion followed by polysaccharide analysis using carbohydrate gel electrophoresis analysis. The results showed that both 1,3-β-D-glucosidic and 1,4-α-D-galactosiduronic linkages were existed in Lingzhi (Ganoderma lucidum and Ganoderma sinense), and the similarity of polysaccharides from G. lucidum and G. sinense was high, which may contribute to rational use of Lingzhi. Different species of Ganoderma and their adulterants can be differentiated based on the saccharide mapping, which is helpful to well understand the structural characters of polysaccharides from different species of Ganoderma and to improve the quality control of polysaccharides in Lingzhi.
Sigma-Aldrich
Pectinase from Aspergillus niger, powder, slightly beige, >1 U/mg
Sigma-Aldrich
Pectinase from Aspergillus niger, BioReagent, suitable for plant cell culture, aqueous glycerol solution, ≥5 units/mg protein (Lowry)
Sigma-Aldrich
Pectolyase from Aspergillus japonicus, suitable for plant cell culture, lyophilized powder
Sigma-Aldrich
Pectinase from Rhizopus sp., powder, 400-800 units/g solid
Sigma-Aldrich
Pectolyase from Aspergillus japonicus, lyophilized powder, ≥0.3 units/mg solid