Merck
  • De novo asymmetric synthesis of cladospolide B-D: structural reassignment of cladospolide D via the synthesis of its enantiomer.
De novo asymmetric synthesis of cladospolide B-D: structural reassignment of cladospolide D via the synthesis of its enantiomer.
Organic letters (2009-02-06)
Yalan Xing, George A O'Doherty
ABSTRACT
The enantioselective synthesis of cladospolide B, C, and (ent)-cladospolide D has been achieved in 11-15 steps from 1-nonyne. The route relies upon an alkyne zipper reaction to relay an ynone and dienoate functional groups across a nine carbon fragment, which enables a highly enantioselective Noyori ynone reduction and a diastereo- and regioselective Sharpless dihydroxylation of a dienoate. In addition to being a flexible approach to three members of the cladospolide natural products, this route for the first time correctly established the structure for cladospolide D.
Sigma-Aldrich
1-Nonyne, 99%