Merck
Childhood thoracic actinomycosis: case report.
Journal of pediatric surgery (2012-06-19)
Alae El Koraichi, Rachid Oulahyane, Asmae Abbassi, Mohamed Y Benjelloun, Mounir Kisra, Najib Benhmamouch
ABSTRACT
Actinomycosis is a rare bacterial infection. It has a pseudotumoral appearance when localized to the lungs or mediastinum. The diagnosis is often delayed because clinical presentation is nonspecific, bacteriologic diagnosis is difficult, and culture growth is slow. The diagnosis is mainly based upon histologic identification of actinomycotic sulfur granules. Actinomycosis is usually sensitive to penicillin G. Surgical treatment is reserved for failure of medical treatment and complications. We report a case of thoracic actinomycosis in a 13-year-old boy.
Product Description
Phenoxymethylpenicillin, European Pharmacopoeia (EP) Reference Standard