Merck
  • High-performance thin-layer chromatography densitometric method for the quantification of harmine, harmaline, vasicine, and vasicinone in Peganum harmala.
High-performance thin-layer chromatography densitometric method for the quantification of harmine, harmaline, vasicine, and vasicinone in Peganum harmala.
Journal of AOAC International (2008-11-05)
Harsha Pulpati, Yogesh S Biradar, Mandapati Rajani
ABSTRACT
Peganum harmala Linn. seed is a reputed drug of the Indian systems of medicine. We report a simple high-performance thin-layer chromatography (HPTLC) densitometric method for the quantification of 4 alkaloids, viz., harmine, harmaline, vasicine, and vasicinone from P. harmala seed. The 4 compounds were resolved in ethyl acetate-methanol-ammonia (7 + 1 + 0.3, v/v/v) mobile phase. The method was validated for precision, repeatability, and accuracy. Instrumental precision was 0.27, 1.53, 0.39, and 1.15% [relative standard deviation (RSD)] and repeatability of the method was 1.01, 0.79, 0.13, and 1.62% RSD for harmine, harmaline, vasicine, and vasicinone, respectively. Accuracy of the method was checked by a recovery study conducted at 3 different levels, and the average recovery was 97.88% for harmine, 97.69% for harmaline, 98.38% for vasicine, and 98.28% for vasicinone. The 4 compounds were quantified in P. harmala seed using the method, and it was found to contain 0.44, 0.096, 0.25, and 0.0007% (w/w) of each, respectively. The proposed HPTLC densitometric method for the quantification of the 4 compounds was found to be simple, precise, specific, sensitive, and accurate. It can be used for routine quality control of P. harmala seed and has the ability to quantify the 4 marker compounds in the formulations containing P. harmala. It also can be used to quantify any of these marker compounds in other herbal drugs.