Merck
Multicomponent synthesis of 3-indolepropionic acids.
Organic letters (2007-01-16)
Mauro F A Adamo, Vivekananda R Konda
ABSTRACT
A three-component one-pot procedure (3-MC) was developed to assemble 3-indolepropionic acids from commercially available materials. This new methodology affords the title compounds in high yields and without the use of chromatography. [reaction: see text].
Sigma-Aldrich
3-Indolepropionic acid, ReagentPlusĀ®, 99%