- [Conductometric method of studying hemocoagulation].
[Conductometric method of studying hemocoagulation].
Meditsinskaia tekhnika (1987-05-01)
V F Rusiaev
PMID3613945
ABSTRACT
A conductometric method for studying flowing blood coagulation is proposed. Phases of conductograms characterizing the kinetics and intensity of blood cells aggregation, release of biologically active compounds, fibrin formation, its mechanical properties, retraction process and fibrinolysis of clot are analysed on the basis of their biophysical mechanisms.