Merck
  • [Postoperative pancreonecrosis prophylaxis in a gastric cancer surgery].
[Postoperative pancreonecrosis prophylaxis in a gastric cancer surgery].
Klinichna khirurhiia (2014-02-08)
Iu A Vinnik, V V Oleksenko, T S Efetova, K A Aliev
ABSTRACT
Efficacy of a new method of intraoperative regional sympathetic blockade of splanchnic nerves was estimated, basing on the results of surgical treatment of 3134 patients, suffering gastric cancer. While comparing the immediate results of surgical interventions there was established, that the risk of postoperative pancreatitis occurrence (lethality) is dependent on the kind of operation performed and its traumaticity. The risk of postoperative pancreonecrosis occurrence (lethality) lowering after standard surgical and combined intervention, after gastric surgery with pancreatic resection was promoted by application of a spiritus-novocaine blockade with the objective to suppress sympathetic reflexes as an element of anesthesia intraoperatively and in early postoperative period.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Procaine hydrochloride, ≥97%
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Procaine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Procaine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications