Merck
Prereplication-complex formation: a molecular double take?
Nature structural & molecular biology (2014-01-07)
Hasan Yardimci, Johannes C Walter
ABSTRACT
In G1, two copies of the MCM2-7 helicase are recruited to each origin of replication. Whereas recruitment of the first MCM2-7 is likely to be analogous to the loading of sliding clamps around DNA, how the second MCM2-7 complex is recruited is highly contentious. Here, we argue that MCM2-7 loading involves specific modifications to the clamp-loading reaction and propose that the first and second MCM2-7 molecules are loaded via similar mechanisms.
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC19, buffered aqueous solution
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from herring testes, Type XIV
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC18, buffered aqueous solution
Sigma-Aldrich
Deoxyribonucleic acid from human placenta, buffered aqueous solution, sexed, female