Merck
  • The properties of an improvised piston pump for the rapid delivery of intravenous fluids.
The properties of an improvised piston pump for the rapid delivery of intravenous fluids.
Anaesthesia (2014-01-05)
C M Smart, C W Primrose, A L Peters, E J Speirits
ABSTRACT
To maximise the effect of a small fluid load, it is occasionally desirable to bolus manually with multiple depressions of a large-capacity syringe. This is usually achieved by placing the syringe on the side port of a three-way tap. We modified this technique by placing two-one-way valves in line with the three-way tap, effectively creating a piston pump, the infusion rates via which we compared with those achieved by an inflatable pressure-infuser in a simulated resuscitation. Fluid flow was faster using the piston pump than with the pressure-infuser (mean (SD) time to infuse 2000 ml saline 0.9% via a 16-G cannula 352 (10) s vs 495 (19) s, respectively, p < 0.0001). The piston pump appears to have potential for both tight control of fluid delivery and major high-volume resuscitation. The lightweight nature of the pump and its lack of reliance on gravity may also make it suitable for the pre-hospital setting.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material