Merck
  • Efficacy and safety of recombinant human bone morphogenetic protein-2/calcium phosphate matrix for closed tibial diaphyseal fracture: a double-blind, randomized, controlled phase-II/III trial.
Efficacy and safety of recombinant human bone morphogenetic protein-2/calcium phosphate matrix for closed tibial diaphyseal fracture: a double-blind, randomized, controlled phase-II/III trial.
The Journal of bone and joint surgery. American volume (2013-12-07)
Thomas Lyon, Wim Scheele, Mohit Bhandari, Kenneth J Koval, Eduardo Gomez Sanchez, Jared Christensen, Alexandre Valentin, Francois Huard
ABSTRACT
Recombinant human bone morphogenetic protein-2 (rhBMP-2) applied on an absorbable collagen sponge improves open tibial fracture-healing as an adjunct to unreamed intramedullary nail fixation. We evaluated rhBMP-2 and a new, injectable calcium phosphate matrix (CPM) formulation in acute closed tibial diaphyseal fractures treated with reamed intramedullary nail fixation. Patients were randomized (1:2:2:1) to receive standard of care, which consisted of definitive fracture fixation within seventy-two hours of injury with a locked intramedullary nail after reaming; standard of care and injection with 1.0 mg/mL of rhBMP-2/CPM; standard of care and injection with 2.0 mg/mL of rhBMP-2/CPM; or standard of care and injection with buffer/CPM, to evaluate the activity of the CPM delivery matrix and provide for sponsor and investigator blinding. The co-primary end points of the study were the effects of rhBMP-2/CPM on the time to fracture union (based on blinded assessment of radiographs) and the time to return to normal function (based on blinded assessment of the time to full weight-bearing without pain at the fracture site) compared with standard of care alone. Three hundred and sixty-nine patients were randomized and included in the intent-to-treat population. This study was terminated after an interim analysis (180 patients with six months of follow-up) revealed no shortening in the time to fracture union in the active treatment arms compared with the standard of care control (the SOC group). In the final primary analysis, the median time to radiographic fracture union was not significantly different for the SOC (13.1 weeks), 1.0-mg/mL rhBMP-2/CPM (13.0 weeks), 2.0-mg/mL rhBMP-2/CPM (15.9 weeks), or buffer/CPM (15.4 weeks) treatment groups. The median time to pain-free full weight-bearing was also not significantly different among the SOC (13.4 weeks), 1.0-mg/mL rhBMP-2/CPM (13.4 weeks), 2.0-mg/mL rhBMP-2/CPM (14.3 weeks), and buffer/CPM (16.4 weeks) treatment groups. In patients with closed tibial fractures treated with reamed intramedullary nailing, the time to fracture union and pain-free full weight-bearing were not significantly reduced by rhBMP-2/CPM compared with standard of care alone. 24306696
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), contains 5 wt. % silica as dopant, synthetic
Sigma-Aldrich
Hydroxyapatite, reagent grade, powder, synthetic
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), ≥97%, synthetic
Sigma-Aldrich
Hydroxyapatite, synthetic, 99.8% trace metals basis (excludes Mg)
Sigma-Aldrich
Calcium phosphate tribasic, 34.0-40.0% Ca basis
Sigma-Aldrich
Calcium phosphate tribasic, suitable for plant cell culture, BioReagent, powder
Sigma-Aldrich
Hydroxyapatite, nanoparticles, dispersion, 10 wt. % in H2O, <200 nm particle size (BET)