Merck
  • Pharmacologic characterization of human male urethral smooth muscle: an in vitro approach.
Pharmacologic characterization of human male urethral smooth muscle: an in vitro approach.
Urology (2013-12-04)
George T Kedia, Matthias Oelke, Michael Sohn, Markus A Kuczyk, Stefan Ückert
ABSTRACT
To elucidate the functional responses of isolated human urethral smooth muscle to various agents known to exert smooth muscle contraction or relaxation. Specimens of penile urethra were obtained from male patients who had undergone male-to-female gender reassignment surgery. Using the tissue bath technique, the contraction induced by increasing concentrations (1 nM-10 μM) of norepinephrine, phenylephrine, acetylcholine, carbachol, prostaglandin F2α, endothelin 1, angiotensin II, and oxytocin was measured. In another set-up, the effects of C-type natriuretic peptide (0.1 nM-1 μM), sodium nitroprusside, sildenafil, forskolin, alpha2-antagonist delquamine, and acetylcholine (1 nM/10 nM-10 μM) on the tension induced by norepinephrine were investigated. The production of cyclic guanosine monophosphate (GMP) and cyclic adenosine monophosphate (AMP) was measured by means of specific radioimmunoassays. Endothelin 1, oxytocin, prostaglandin F2α, norepinephrine, and phenylephrine induced dose-dependent contraction of the isolated urethral tissue, whereas acetylcholine, carbachol, and angiotensin II had no or only minor contractile effects. The contraction induced by norepinephrine was reversed by the drugs with the following rank order of efficacy: sodium nitroprusside > delquamine > sildenafil > C-type natriuretic peptide > forskolin > acetylcholine. The maximal reversion of tension ranged from 68% (sodium nitroprusside) to 22% (acetylcholine). The relaxing effects of the drugs were paralleled by a several-fold increase in tissue levels of cyclic GMP and cyclic adenosine monophosphate. The results provide evidence that urethral smooth muscle is under the control of endogenous compounds, such as adrenergic agonists (norepinephrine and phenylephrine), vasoactive peptides, prostagladins, NO/cyclic GMP, and acetylcholine, assumed to influence micturition at the peripheral level.
Product Description
Supelco
Phenylephrine Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Norepinephrine hydrochloride, ≥98.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate sodium salt monohydrate, ≥98.0% (HPLC), powder
Sigma-Aldrich
DL-Norepinephrine hydrochloride, crystalline, ≥97% (TLC)
Sigma-Aldrich
(−)-Norepinephrine, ≥98%, crystalline
Sigma-Aldrich
(±)-Norepinephrine (+)-bitartrate salt
Sigma-Aldrich
Oxytocin acetate salt hydrate, ≥97% (HPLC)
Sigma-Aldrich
Guanosine 3′,5′-cyclic monophosphate, ≥98% (HPLC), powder
Sigma-Aldrich
Oxytocin
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, analytical standard
Sigma-Aldrich
Oxytocin, lyophilized powder, ~15 IU/mg solid (Prepared from synthetic oxytocin)
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, powder
Phenylephrine hydrochloride for peak identification, European Pharmacopoeia (EP) Reference Standard
Phenylephrine, European Pharmacopoeia (EP) Reference Standard
Oxytocin, European Pharmacopoeia (EP) Reference Standard