Merck
  • Impact of addition of aromatic amino acids on non-volatile and volatile compounds in lychee wine fermented with Saccharomyces cerevisiae MERIT.ferm.
Impact of addition of aromatic amino acids on non-volatile and volatile compounds in lychee wine fermented with Saccharomyces cerevisiae MERIT.ferm.
International journal of food microbiology (2013-11-30)
Dai Chen, Jing Yee Chia, Shao-Quan Liu
ABSTRACT
The impact of individual aromatic amino acid addition (L-phenylalanine, L-tryptophan and L-tyrosine) on non-volatile and volatile constituents in lychee wine fermented with Saccharomyces cerevisiae var. cerevisiae MERIT.ferm was studied. None of the added amino acids had any significant effect on the yeast cell count, pH, soluble solid contents, sugars and ethanol. The addition of L-phenylalanine significantly reduced the production of pyruvic and succinic acids. The addition of each amino acid dramatically reduced the consumption of proline and decreased the production of glycerol. Supplementation of the lychee juice with L-phenylalanine resulted in the formation of significantly higher amounts of 2-phenylethyl alcohol, 2-phenylethyl acetate, 2-phenylethyl isobutyrate and 2-phenylethyl hexanoate. In contrast, supplementation with L-tryptophan and L-tyrosine had negligible effects on the volatile profile of lychee wines. These findings suggest that selectively adding amino acids may be used as a tool to modulate the volatile profile of lychee wines so as to diversify and/or intensify wine flavour and style.
Sigma-Aldrich
Phenethyl acetate, natural, ≥98%, FCC, FG
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Phenethyl acetate, ≥98%, FCC, FG
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Phenylethyl Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Phenethyl acetate, 99%
Sigma-Aldrich
Phenethyl alcohol, natural, ≥99%, FCC, FG
Sigma-Aldrich
Phenethyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
2-Phenylethanol, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)