Merck
  • A metabolic shift favoring sphingosine 1-phosphate at the expense of ceramide controls glioblastoma angiogenesis.
A metabolic shift favoring sphingosine 1-phosphate at the expense of ceramide controls glioblastoma angiogenesis.
The Journal of biological chemistry (2013-11-23)
Hazem J Abuhusain, Azadeh Matin, Qiao Qiao, Han Shen, Nupur Kain, Bryan W Day, Brett W Stringer, Benjamin Daniels, Maarit A Laaksonen, Charlie Teo, Kerrie L McDonald, Anthony S Don
ABSTRACT
Studies in cell culture and mouse models of cancer have indicated that the soluble sphingolipid metabolite sphingosine 1-phosphate (S1P) promotes cancer cell proliferation, survival, invasiveness, and tumor angiogenesis. In contrast, its metabolic precursor ceramide is prodifferentiative and proapoptotic. To determine whether sphingolipid balance plays a significant role in glioma malignancy, we undertook a comprehensive analysis of sphingolipid metabolites in human glioma and normal gray matter tissue specimens. We demonstrate, for the first time, a systematic shift in sphingolipid metabolism favoring S1P over ceramide, which increases with increasing cancer grade. S1P content was, on average, 9-fold higher in glioblastoma tissues compared with normal gray matter, whereas the most abundant form of ceramide in the brain, C18 ceramide, was on average 5-fold lower. Increased S1P content in the tumors was significantly correlated with increased sphingosine kinase 1 (SPHK1) and decreased sphingosine phosphate phosphatase 2 (SGPP2) expression. Inhibition of S1P production by cultured glioblastoma cells, using a highly potent and selective SPHK1 inhibitor, blocked angiogenesis in cocultured endothelial cells without affecting VEGF secretion. Our findings validate the hypothesis that an altered ceramide/S1P balance is an important feature of human cancers and support the development of SPHK1 inhibitors as antiangiogenic agents for cancer therapy.
Sigma-Aldrich
D-Sphingosine, synthetic
SKU
Pack Size
Availability
Price
Quantity
Avanti
Sphingosine (d17:1), Avanti Research - A Croda Brand LM2000, ethanol solution
SKU
Pack Size
Availability
Price
Quantity
Avanti
Sphingosine-1-Phosphate (d18:1), Avanti Research - A Croda Brand
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sphingosine 1-phosphate, ≥95%, powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sphingosine 1-phosphate, ≥98.0% (TLC)
SKU
Pack Size
Availability
Price
Quantity
Avanti
17:0-20:4 PI(3,4,5)P3, Avanti Research - A Croda Brand LM1906, powder