Merck
  • Differences in gastrointestinal calcium absorption after the ingestion of calcium-free phosphate binders.
Differences in gastrointestinal calcium absorption after the ingestion of calcium-free phosphate binders.
American journal of physiology. Renal physiology (2013-11-08)
Geert J Behets, Geert Dams, Stephen J Damment, Patrick Martin, Marc E De Broe, Patrick C D'Haese
ABSTRACT
Both calcium-containing and noncalcium-containing phosphate binders can increase gastrointestinal calcium absorption. Previously, we observed that lanthanum carbonate administration to rats with renal failure is not associated with increased calciuria. Additionally, lanthanum carbonate treatment in dialysis patients has been associated with a less pronounced initial decrease in serum parathyroid hormone compared with other phosphate binders. For 8 days, male Wistar rats received a diet supplemented with 2% lanthanum carbonate, 2% sevelamer, 2% calcium carbonate, or 2% cellulose. Calciuria was found to be increased in animals with normal renal function treated with sevelamer or calcium carbonate but not with lanthanum carbonate. In animals with renal failure, cumulative calcium excretion showed similar results. In rats with normal renal function, serum ionized calcium levels were increased after 2 days of treatment with sevelamer, while calcium carbonate showed a smaller increase. Lanthanum carbonate did not induce differences. In animals with renal failure, no differences were found between sevelamer-treated, calcium carbonate-treated, and control groups. Lanthanum carbonate, however, induced lower ionized calcium levels within 2 days of treatment. These results were confirmed in normal human volunteers, who showed lower net calcium absorption after a single dose of lanthanum carbonate compared with sevelamer carbonate. In conclusion, these two noncalcium-containing phosphate-binding agents showed a differential effect on gastrointestinal calcium absorption. These findings may help to improve the management of calcium balance in patients with renal failure, including concomitant use of vitamin D.
Product Description
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
α-Cellulose, powder
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Cellulose, fibers, (medium)
Supelco
Cellulose, powder, for column chromatography
Supelco
Cellulose, DS-0, powder, suitable for thin layer chromatography (TLC)
Supelco
Cellulose, acid washed, powder, for column chromatography
Supelco
Ammonium carbamate, for decomposition
Supelco
Cellulose, acid washed, from spruce, for column chromatography
Sigma-Aldrich
Cellulose, microcrystalline, powder
Sigma-Aldrich
Lanthanum, pieces
Sigma-Aldrich
Cellulose, colloidal, microcrystalline
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.
Supelco
Cellulose, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Sigma-Aldrich
Cellulose, microcrystalline, powder, 20 μm
Sigma-Aldrich
Lanthanum, powder, −40 mesh, under oil, 99.9% trace rare earth metals basis
Sigma-Aldrich
Ammonium carbamate, 99%