Merck
  • Treatment with thyroxine restores myelination and clinical recovery after intraventricular hemorrhage.
Treatment with thyroxine restores myelination and clinical recovery after intraventricular hemorrhage.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2013-11-01)
Linnea R Vose, Govindaiah Vinukonda, Sungro Jo, Omid Miry, Daniel Diamond, Ritesh Korumilli, Arslan Arshad, Muhammad T K Zia, Furong Hu, Robert J Kayton, Edmund F La Gamma, Rashmi Bansal, Antonio C Bianco, Praveen Ballabh
ABSTRACT
Intraventricular hemorrhage (IVH) remains a major cause of white matter injury in preterm infants with no viable therapeutic strategy to restore myelination. Maturation of oligodendrocytes and myelination is influenced by thyroid hormone (TH) signaling, which is mediated by TH receptor α (TRα) and TRβ. In the brain, cellular levels of TH are regulated by deiodinases, with deiodinase-2 mediating TH activation and deiodinase-3 TH inactivation. Therefore, we hypothesized that IVH would decrease TH signaling via changes in the expression of deiodinases and/or TRs, and normalization of TH signaling would enhance maturation of oligodendrocytes and myelination in preterm infants with IVH. These hypotheses were tested using both autopsy materials from human preterm infants and a rabbit model of IVH. We found that deiodinase-2 levels were reduced, whereas deiodinase-3 levels were increased in brain samples of both humans and rabbits with IVH compared with controls without IVH. TRα expression was also increased in human infants with IVH. Importantly, treatment with TH accelerated the proliferation and maturation of oligodendrocytes, increased transcription of Olig2 and Sox10 genes, augmented myelination, and restored neurological function in pups with IVH. Consistent with these findings, the density of myelinating oligodendrocytes was almost doubled in TH-treated human preterm infants compared with controls. Thus, in infants with IVH the combined elevation in deiodinase-3 and reduction in deiodinase-2 decreases TH signaling that can be worsened by an increase in unliganded TRα. Given that TH promotes neurological recovery in IVH, TH treatment might improve the neurodevelopmental outcome of preterm infants with IVH.
Sigma-Aldrich
L-Thyroxine, ≥98% (HPLC)
Sigma-Aldrich
L-Thyroxine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
ApopTag Fluorescein In Situ Apoptosis Detection Kit, The ApopTag Fluorescein In Situ Apoptosis Detection Kit detects apoptotic cells in situ by the indirect TUNEL method, utilizing an anti-digoxigenin antibody that is conjugated to a Fluorescein reporter molecule.