Merck
Asterixis related to gabapentin intake: a case report and review.
Postgraduate medicine (2013-10-12)
Mervat Wahba, Olga Waln
ABSTRACT
Asterixis has been reported as a side effect of toxic and metabolic abnormalities. The current literature contains very little evidence of gabapentin-related neurotoxicity. Our observations reveal that neurotoxic side effects of gabapentin use may be seen even when dose adjustments are made for patients with mild-to-moderate renal dysfunction. Clinical vigilance for possible neurotoxic effects of gabapentin therapy in patients with renal dysfunction is warranted.
Product Description
Supelco
Gabapentin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Gabapentin Related Compound A, Pharmaceutical Secondary Standard; Certified Reference Material