Merck
  • Potentiated macrophage activation by acid sensing under low adiponectin levels.
Potentiated macrophage activation by acid sensing under low adiponectin levels.
Molecular immunology (2013-10-03)
Takaharu Negoro, Masaoki Kin, Akitoshi Takuma, Kiyomi Saito, Shunichi Shimizu, Yasuko Nakano
ABSTRACT
Adiponectin can protect against inflammation; one of the mechanisms involves direct, inhibition of macrophages (MΦ). We postulated that adiponectin anti-sense transgenic (AsTg) mice raised in our laboratory are prone to inflammation because of systemic low adiponectin levels. The writhing response to acetic acid was utilized as an in vivo inflammatory model, and using Ca(2)(+), response to the acid was exploited in vitro to evaluate the function of resident peritoneal MΦ. The in vivo response to the acid was increased and the Ca(2)(+) response of MΦ was enhanced in AsTg mice, compared with those in wild type (WT) mice. In parallel with these enhanced responses, MΦ from AsTg mice augmented TNF-α and IL-6 mRNA expression. We further analyzed the enhancement in activity of MΦ from AsTg mice by acid sensing using specific inhibitors, amiloride for acid-sensing ion channels (ASICs) and KB-R7943 for Na(+)/Ca(2)(+) exchangers (NCXs). Our results indicated that in AsTg mice, the Ca(2)(+) response to the acid was facilitated in MΦ by a low threshold of ASIC1 and NCX1 molecules and the activity of these channel was possibly regulated by adiponectin.
Product Description
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
USP
Amiloride hydrochloride, United States Pharmacopeia (USP) Reference Standard
Millipore
Bifido Selective Supplement B, suitable for microbiology