Merck
  • CTP synthase 1, a smooth muscle-sensitive therapeutic target for effective vascular repair.
CTP synthase 1, a smooth muscle-sensitive therapeutic target for effective vascular repair.
Arteriosclerosis, thrombosis, and vascular biology (2013-09-07)
Rui Tang, Xiao-Bing Cui, Jia-Ning Wang, Shi-You Chen
ABSTRACT
Vascular remodeling as a result of smooth muscle cell (SMC) proliferation and neointima formation is a major medical challenge in cardiovascular intervention. However, antineointima drugs often indistinguishably block re-endothelialization, an essential step toward successful vascular repair, because of their nonspecific effect on endothelial cells (ECs). The objective of this study is to identify a therapeutic target that differentially regulates SMC and EC proliferation. Using both rat balloon injury and mouse wire injury models, we identified CTP synthase 1 (CTPS1) as one of the potential targets that may be used for developing therapeutics for treating neointima-related disorders. CTPS1 was induced in proliferative SMCs in vitro and neointima SMCs in vivo. Blockade of CTPS1 expression by small hairpin RNA or activity by cyclopentenyl cytosine suppressed SMC proliferation and neointima formation. Surprisingly, cyclopentenyl cytosine had much less effect on EC proliferation. Of importance, blockade of CTPS1 in vivo sustained the re-endothelialization as a result of induction of CTP synthesis salvage pathway enzymes nucleoside-diphosphate kinase A and B in ECs. Diphosphate kinase B seemed to preserve EC proliferation via use of extracellular cytidine to synthesize CTP. Indeed, blockade of both CTPS1 and diphosphate kinase B suppressed EC proliferation in vitro and the re-endothelialization in vivo. Our study uncovered a fundamental difference in CTP biosynthesis between SMCs and ECs during vascular remodeling, which provided a novel strategy by using cyclopentenyl cytosine or other CTPS1 inhibitors to selectively block SMC proliferation without disturbing or even promoting re-endothelialization for effective vascular repair after injury.
Sigma-Aldrich
Cytidine, 99%
Sigma-Aldrich
Cytidine, BioReagent, suitable for cell culture, powder, ≥99%