Merck
  • Cloning, characterization, and inhibition studies of a β-carbonic anhydrase from Leishmania donovani chagasi, the protozoan parasite responsible for leishmaniasis.
Cloning, characterization, and inhibition studies of a β-carbonic anhydrase from Leishmania donovani chagasi, the protozoan parasite responsible for leishmaniasis.
Journal of medicinal chemistry (2013-08-28)
Leo Syrjänen, Alane Beatriz Vermelho, Igor de Almeida Rodrigues, Suzana Corte-Real, Terhi Salonen, Peiwen Pan, Daniela Vullo, Seppo Parkkila, Clemente Capasso, Claudiu T Supuran
ABSTRACT
Leishmaniasis is an infection provoked by protozoans belonging to the genus Leishmania. Among the many species and subsepecies of such protozoa, Leishmania donovani chagasi causes visceral leishmaniasis. A β-carbonic anhydrase (CA, EC 4.2.1.1) was cloned and characterized from this organism, denominated here LdcCA. LdcCA possesses effective catalytic activity for the CO2 hydration reaction, with kcat of 9.35 × 10(5) s(-1) and kcat/KM of 5.9 × 10(7) M(-1) s(-1). A large number of aromatic/heterocyclic sulfonamides and 5-mercapto-1,3,4-thiadiazoles were investigated as LdcCA inhibitors. The sulfonamides were medium potency to weak inhibitors (KI values of 50.2 nM-9.25 μM), whereas some heterocyclic thiols inhibited the enzyme with KIs in the range of 13.4-152 nM. Some of the investigated thiols efficiently inhibited the in vivo growth of Leishmania chagasi and Leishmania amazonensis promastigotes, by impairing the flagellar pocket and movement of the parasites and causing their death. The β-CA from Leishmania spp. is proposed here as a new antileishmanial drug target.
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II human, ≥80%, powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, Isoelectric focusing marker, pI 5.4
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, ≥95% (SDS-PAGE), specific activity ≥3,500 W-A units/mg protein, lyophilized powder
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, BioReagent, suitable for GFC marker
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, non-denaturing PAGE marker
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, For use as a marker in SDS-PAGE
Sigma-Aldrich
Carbonic Anhydrase I from human erythrocytes
Supelco
Carbonic Anhydrase I from human erythrocytes, Isoelectric focusing marker, pI 6.6