Merck
  • Isotope labeling studies on the electron impact mass spectral fragmentation patterns of chloropropanol acetates.
Isotope labeling studies on the electron impact mass spectral fragmentation patterns of chloropropanol acetates.
Journal of agricultural and food chemistry (2013-08-24)
Anja K K Rahn, Varoujan A Yaylayan
ABSTRACT
Chloropropanol (CP) esters are part of an emerging group of process-induced toxicants that are considered as potential health hazards particularly in palm oil. Mass spectrometry-based methodologies for identification of CP esters in food are critical in overcoming the challenges associated with direct detection methods. In the present study, a convenient strategy was employed to generate all possible CP acetates through reacting acetic anhydride with either glycerol in the presence of a chloride source or the corresponding CPs, such as 3-chloro-, 1,3-dichloro-, 2-chloro-, and 1,2-dichloropropanols, allowing for the identification of the individual CP acetates and assignment of their mass spectral fragmentations. Mass spectral fragmentations were confirmed through the use of the isotopic signature of chlorine in addition to the isotope labeling experiments performed using isotopically labeled precursors, such as [¹³C-U₃] glycerol, [¹³C-U₄] acetic anhydride, [¹³C-2,2'] acetic anhydride, and [d₅] 3-monochloropropane-1,2-diol (3-MCPD) as reactants. Such studies have indicated that all CP esters can undergo two general fragmentations under electron impact (EI) conditions, one generating the acylium ion at m/z 45 and the other generating a chlorinated cyclic acyloxonium ion at m/z 135.6. Considering the fact that such ions can also be generated from any fatty acid containing CP esters after undergoing McLafferty rearrangement, the ion at m/z 135.6 can therefore be considered as a universal marker for the presence of CP esters undergoing EI fragmentation. Furthermore, these studies have also indicated the formation of ions characteristic of CP diesters, monoesters, and dichloro esters.
Product Description
Supelco
1-Propanol, analytical standard
Sigma-Aldrich
1-Propanol, anhydrous, 99.7%
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
1-Propanol, ≥99% (GC), purum
Sigma-Aldrich
1-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
1-Propanol, suitable for HPLC, ≥99.9%