Merck
Comparative toxicity of CuO nanoparticles and CuSO4 in rainbow trout.
Ecotoxicology and environmental safety (2013-08-13)
Gloria Isani, Maria Letizia Falcioni, Gianni Barucca, Durairaj Sekar, Giulia Andreani, Emilio Carpenè, Giancarlo Falcioni
ABSTRACT
This study compared the toxicity and accumulation of two different Cu compounds, CuO nanoparticles (NPs) and soluble CuSO4, in erythrocytes and different tissues in rainbow trout (Oncorhynchus mykiss). The crystal structure of CuO NP analysed by XRD indicates that the NP are Tenorite, a monoclinic CuO. The in vitro toxicity results indicate that both Cu compounds increase the haemolysis rate in a dose-dependent way, but the effect was reduced treating cells with CuO NP. Moreover, both Cu compounds induce DNA damage and the entity of the damage, similarly to haemolysis, was more marked in cells treated with CuSO4. In vivo results, obtained after intraperitoneal injection, showed that Cu concentrations were significantly higher in gills (p<0.0001), kidney (p=0.007) and liver (p<0.05) of exposed fish with a significant increase in plasma Cu concentration 15h after CuSO4 treatment. Cu concentrations were significantly higher in fish exposed to CuSO4 than CuO in kidney (p<0.05) and gills (p<0.0001). Significant DNA damage with respect to controls was detected only when Cu was injected as CuSO4. The present data could serve to evaluate environmental Cu toxicity in fish depending on Cu speciation.
Sigma-Aldrich
Copper(II) oxide, powder, 99.995% trace metals basis
Sigma-Aldrich
Copper(II) oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Copper(II) sulfate, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Copper(II) oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Copper(II) oxide, 99.999% trace metals basis
Sigma-Aldrich
Copper(II) sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99-100.5% (based on anhydrous substance)
Sigma-Aldrich
Copper(II) oxide, powder, <10 μm, 98%
Sigma-Aldrich
Copper(II) sulfate, puriss. p.a., anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Copper(II) sulfate solution, 4 % (w/v) (prepared from copper (II) sulfate pentahydrate)
Sigma-Aldrich
Copper(II) oxide, needles, mixture of CuO and Cu2O, ACS reagent
Sigma-Aldrich
Copper(II) sulfate, ReagentPlus®, ≥99%
Sigma-Aldrich
Copper(II) oxide on alumina, 14-30 mesh, extent of labeling: 13 wt. % loading
Sigma-Aldrich
Copper(II) oxide, ACS reagent, ≥99.0%