Merck
  • Biomimetic poly(glycerol sebacate) (PGS) membranes for cardiac patch application.
Biomimetic poly(glycerol sebacate) (PGS) membranes for cardiac patch application.
Materials science & engineering. C, Materials for biological applications (2013-08-06)
Ranjana Rai, Marwa Tallawi, Niccoletta Barbani, Caterina Frati, Denise Madeddu, Stefano Cavalli, Gallia Graiani, Federico Quaini, Judith A Roether, Dirk W Schubert, Elisabetta Rosellini, Aldo R Boccaccini
ABSTRACT
In this study biomimetic poly(glycerol sebacate) PGS matrix was developed for cardiac patch application. The rationale was that such matrices would provide conducive environment for the seeded cells at the interphase with PGS. From the microstructural standpoint, PGS was fabricated into dense films and porous PGS scaffolds. From the biological aspect, biomimetic PGS membranes were developed via covalently binding peptides Tyr-Ile-Gly-Ser-Arg (YIGSR) and Gly-Arg-Gly-Asp-Ser-Pro (GRGDSP), corresponding to the epitope sequences of laminin and fibronectin, respectively onto the surface. To improve and enhance homogenous binding of peptides onto the PGS surface, chemical modification of its surface was carried out. A sequential regime of alkaline hydrolysis with 0.01 M NaOH for 5 min and acidification with 0.01 M HCl for 25s was optimal. More COOH chemical group was exposed without causing deleterious effect on the bulk properties of the polymer as revealed by the physicochemical analysis carried out. HPLC analysis, chemical imaging and ToF-SIMS were able to establish the successful homogenous functionalization of PGS membranes with the peptides. Finally, the developed biomimetic membranes supported the adhesion and growth of rat and human cardiac progenitor cells.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Ascarite®, Sodium hydroxide-coated silica, 20-30 mesh