Merck
  • Facile synthesis of low crystalline MoS2 nanosheet-coated CNTs for enhanced hydrogen evolution reaction.
Facile synthesis of low crystalline MoS2 nanosheet-coated CNTs for enhanced hydrogen evolution reaction.
Nanoscale (2013-07-26)
Ya Yan, Xiaoming Ge, Zhaolin Liu, Jing-Yuan Wang, Jong-Min Lee, Xin Wang
ABSTRACT
In this work, a networked MoS2/CNT nanocomposite has been synthesized by a facile solvothermal method. The as-prepared sample exhibits high catalytic activity for electrocatalytic hydrogen evolution.
Sigma-Aldrich
Molybdenum, powder, 1-5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, nanopowder, <100 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 10 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Molybdenum(IV) sulfide, powder
Sigma-Aldrich
Molybdenum(IV) sulfide, powder, <2 μm, 98%
Sigma-Aldrich
Molybdenum, wire, diam. 1.0 mm, 99.95% trace metals basis