Merck
  • Oral antimycobacterial therapy in chronic cutaneous sarcoidosis: a randomized, single-masked, placebo-controlled study.
Oral antimycobacterial therapy in chronic cutaneous sarcoidosis: a randomized, single-masked, placebo-controlled study.
JAMA dermatology (2013-07-19)
Wonder P Drake, Kyra Oswald-Richter, Bradley W Richmond, Joan Isom, Victoria E Burke, Holly Algood, Nicole Braun, Thyneice Taylor, Kusum V Pandit, Caroline Aboud, Chang Yu, Naftali Kaminski, Alan S Boyd, Lloyd E King
ABSTRACT
Sarcoidosis is a chronic granulomatous disease for which there are limited therapeutic options. This is the first randomized, placebo-controlled study to demonstrate that antimycobacterial therapy reduces lesion diameter and disease severity among patients with chronic cutaneous sarcoidosis. To evaluate the safety and efficacy of once-daily antimycobacterial therapy on the resolution of chronic cutaneous sarcoidosis lesions. A randomized, placebo-controlled, single-masked trial on 30 patients with symptomatic chronic cutaneous sarcoidosis lesions deemed to require therapeutic intervention. A tertiary referral dermatology center in Nashville, Tennessee. Participants were randomized to receive either the oral concomitant levofloxacin, ethambutol, azithromycin, and rifampin (CLEAR) regimen or a comparative placebo regimen for 8 weeks with a 180-day follow-up. Participants were monitored for absolute change in lesion diameter and decrease in granuloma burden, if present, on completion of therapy. In the intention-to-treat analysis, the CLEAR-treated group had a mean (SD) decrease in lesion diameter of -8.4 (14.0) mm compared with an increase of 0.07 (3.2) mm in the placebo-treated group (P = .05). The CLEAR group had a significant reduction in granuloma burden and experienced a mean (SD) decline of -2.9 (2.5) mm in lesion severity compared with a decline of -0.6 (2.1) mm in the placebo group (P = .02). Antimycobacterial therapy may result in significant reductions in chronic cutaneous sarcoidosis lesion diameter compared with placebo. These observed reductions, associated with a clinically significant improvement in symptoms, were present at the 180-day follow-up period. Transcriptome analysis of sarcoidosis CD4+ T cells revealed reversal of pathways associated with disease severity and enhanced T-cell function following T-cell receptor stimulation. clinicaltrials.gov Identifier: NCT01074554.
Product Description
Supelco
Azithromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Azithromycin for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethambutol dihydrochloride, antimycobacterial
Azithromycin, European Pharmacopoeia (EP) Reference Standard
Azithromycin for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Azithromycin dihydrate, ≥98% (HPLC)
Supelco
Ofloxacin, VETRANAL®, analytical standard
Supelco
Ofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Azithromycin
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Supelco
Levofloxacin, analytical standard