Merck
  • Micafungin versus amphotericin B lipid complex for the prevention of invasive fungal infections in high-risk liver transplant recipients.
Micafungin versus amphotericin B lipid complex for the prevention of invasive fungal infections in high-risk liver transplant recipients.
Transplantation (2013-07-12)
Hsin-Yun Sun, Thomas V Cacciarelli, Nina Singh
ABSTRACT
Limited data exist regarding echinocandins as antifungal prophylaxis in liver transplant recipients. The efficacy and safety of targeted prophylaxis with micafungin or amphotericin B lipid complex (ABLC) was assessed in a sequential cohort of high-risk patients (posttransplantation dialysis, retransplantation, or reoperation) and compared with those without high risk who did not receive prophylaxis. Outcomes were assessed at 90 days. Micafungin versus ABLC recipients were older (P=0.0065) and more likely to have hepatocellular carcinoma (P=0.025). High-risks, that is, dialysis (55.6% vs. 79.2%), retransplantation (5.6% vs. 12.5%), and reoperation (38.9% vs. 20.8%) did not differ between the two groups. Invasive fungal infections developed in 11.1% (2 of 18) of micafungin recipients, 8.3% (2 of 24) of ABLC recipients, and 3% (7 of 234) of patients without high risks (P=0.12). In nondialyzed patients, ABLC versus micafungin recipients had significantly higher serum creatinine on day 14 (P=0.04). However, renal and hepatic function, rejection, graft loss, and mortality did not differ for the two groups on day 90. Targeted prophylaxis with micafungin or ABLC decreased the risk of mycoses in high-risk recipients compared with that in low-risk recipients. Compared with ABLC, however, micafungin appeared to be associated with lower early-renal dysfunction and no additional risk of hepatic dysfunction.
Sigma-Aldrich
Amphotericin B solubilized, powder, γ-irradiated, BioXtra, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Amphotericin B from Streptomyces sp., ~80% (HPLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Amphotericin B from Streptomyces sp., BioReagent, suitable for cell culture, ~80% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Amphotericin B for peak identification, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.