Merck
  • Transition polarizability model of induced resonance Raman optical activity.
Transition polarizability model of induced resonance Raman optical activity.
Journal of computational chemistry (2013-07-05)
Shigeki Yamamoto, Petr Bouř
ABSTRACT
Induced resonance Raman optical activity (IRROA) proved to be a very sensitive method to detect molecular chirality. It is exhibited, for example, by complexes of lanthanides with chiral alcohols or ketones. So far, the phenomenon has not been understood at a quantitative level. To elucidate its mechanisms and to correctly relate the spectra to the structure, a transition polarizability model (TPM) is developed and applied to a camphor-europium complex. The model well reproduces the high ROA/Raman intensity ratio of the IRROA observed experimentally. The results additionally indicate a fundamental role of the nonchiral fod ligand in the Eu(fod)3 compound for the chirality enhancement. The TPM model thus serves as a guidance for both experimental and theoretical studies to come.
Sigma-Aldrich
(±)-Camphor, ≥95.5%
Sigma-Aldrich
(1R)-(+)-Camphor, 98%
Sigma-Aldrich
(±)-Camphor, purum, synthetic, ≥95.0% (GC)
Supelco
(−)-Camphor, analytical standard
Sigma-Aldrich
(±)-Camphor, meets analytical specification of Ph. Eur., BP, ≥95% (GC), racemic
Sigma-Aldrich
Camphor, 96%
Camphor (racemic), European Pharmacopoeia (EP) Reference Standard
Camphor (dl), primary reference standard
Sigma-Aldrich
(1S)-(−)-Camphor, 95%
Sigma-Aldrich
D-Camphor, ≥97%, FG
Supelco
D-Camphor, Pharmaceutical Secondary Standard; Certified Reference Material